The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108715.1 | NP_001102185.1 | 9 - 9 | 2921 | 129 |
XM_006240286.2 | XP_006240348.1 | 9 - 9 | 3269 | 129 | |
XM_006240287.2 | XP_006240349.1 | 9 - 9 | 3104 | 129 | |
GenBank mRNA | BC166432.1 | - | 9 - 9 | 3085 | 129 |