Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
Wdr92
Entrez Gene ID
498418
Gene Name
WD repeat domain 92
Gene Aliases
RGD1563863
Chromosome Location
Chr.14: 100373129 - 100394143 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127559.2 NP_001121031.1 3 - 3 452 155
GenBank mRNA BC162042.1 - 3 - 3 452 155

View Full Product Details