The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109092.1 | NP_001102562.1 | 8 - 8 | 1531 | 129 |
XM_006251468.1 | XP_006251530.1 | 8 - 8 | 1526 | 129 | |
XM_006251469.2 | XP_006251531.1 | 8 - 8 | 1730 | 129 | |
XM_006251470.2 | XP_006251532.1 | 7 - 7 | 1402 | 129 |