The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011913.1 | NP_001011913.1 | 3 - 3 | 2786 | 151 |
XM_006251476.2 | XP_006251538.1 | 3 - 3 | 2809 | 151 | |
XM_006251477.2 | XP_006251539.1 | 3 - 3 | 3019 | 151 | |
XM_008770261.1 | XP_008768483.1 | 3 - 3 | 2833 | 151 | |
GenBank mRNA | AY623031.1 | - | 3 - 3 | 2786 | 151 |
AY623032.1 | - | 3 - 3 | 2738 | 151 |