The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134962.1 | NP_001128434.1 | 15 - 15 | 3105 | 85 |
XM_008759532.1 | XP_008757754.1 | 15 - 15 | 3105 | 85 | |
XM_008759533.1 | XP_008757755.1 | 14 - 14 | 2713 | 85 | |
XM_008759534.1 | XP_008757756.1 | 15 - 15 | 2478 | 85 | |
XM_008759535.1 | XP_008757757.1 | 13 - 13 | 2445 | 85 | |
GenBank mRNA | BC169015.1 | - | 15 - 15 | 3105 | 85 |