Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mex3a
Entrez Gene ID
310631
Gene Name
mex-3 RNA binding family member A
Gene Aliases
RGD1564904
Chromosome Location
Chr.2: 187893147 - 187902546 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006232798.2 XP_006232860.2 3 - 3 5400 84

View Full Product Details