Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
Da2-19
Entrez Gene ID
302729
Gene Name
uncharacterized LOC302729
Gene Aliases
-
Chromosome Location
Chr.X: 48196789 - 48213506 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY325254.1 - 3 - 3 129 141

View Full Product Details