The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022800.1 | NP_073637.1 | 2 - 2 | 1185 | 78 |
XM_006232406.2 | XP_006232468.1 | 3 - 3 | 1471 | 78 | |
XM_006232407.2 | XP_006232469.1 | 3 - 3 | 1561 | 78 | |
XM_006232408.2 | XP_006232470.1 | 3 - 3 | 2707 | 78 | |
GenBank mRNA | AF313450.1 | - | 2 - 2 | 1185 | 78 |