Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krt2
Entrez Gene ID
406228
Gene Name
keratin 2, type II
Gene Aliases
Kb2
Chromosome Location
Chr.7: 143412858 - 143420463 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008899.1 NP_001008899.1 8 - 9 1562 83
XM_006242436.2 XP_006242498.1 8 - 9 2063 83

View Full Product Details