Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Armc5
Entrez Gene ID
361653
Gene Name
armadillo repeat containing 5
Gene Aliases
-
Chromosome Location
Chr.1: 199655647 - 199662414 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009455.1 NP_001009455.1 5 - 6 2198 165
GenBank mRNA BC087086.1 - 5 - 6 2198 165

View Full Product Details