The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009486.1 | NP_001009486.1 | 3 - 4 | 283 | 152 |
GenBank mRNA | AY649835.1 | - | 3 - 4 | 283 | 152 |