Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Thrap3
Entrez Gene ID
313591
Gene Name
thyroid hormone receptor associated protein 3
Gene Aliases
-
Chromosome Location
Chr.5: 144178248 - 144221317 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009693.1 NP_001009693.1 9 - 10 2459 145
XM_006238866.2 XP_006238928.1 9 - 10 2439 145
XM_006238867.1 XP_006238929.1 9 - 10 2000 145
XM_008764021.1 XP_008762243.1 10 - 11 2558 145
XM_008764022.1 XP_008762244.1 9 - 10 2808 145
GenBank mRNA BC088415.1 - 9 - 10 2459 145

View Full Product Details