The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024374.4 | NP_077350.1 | 2 - 3 | 305 | 150 |
GenBank mRNA | AY951952.1 | - | - | 312 | 150 |
BC088136.1 | - | 2 - 3 | 305 | 150 | |
D26179.1 | - | 2 - 3 | 360 | 150 | |
FQ228349.1 | - | - | 426 | 150 | |
U21661.1 | - | 2 - 3 | 384 | 150 |