Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Usf1
Entrez Gene ID
83586
Gene Name
upstream transcription factor 1
Gene Aliases
-
Chromosome Location
Chr.13: 89797750 - 89805561 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031777.2 NP_113965.1 7 - 8 690 145
XM_006250266.2 XP_006250328.1 7 - 8 875 145
XM_008769765.1 XP_008767987.1 5 - 6 526 145
GenBank mRNA AF026476.1 - 5 - 6 559 145
BC088849.1 - 7 - 8 690 145

View Full Product Details