Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cxadr
Entrez Gene ID
89843
Gene Name
coxsackie virus and adenovirus receptor
Gene Aliases
Car1
Chromosome Location
Chr.11: 16826298 - 16873564 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053570.1 NP_446022.1 4 - 5 638 171
XM_006248000.2 XP_006248062.1 4 - 5 739 171
GenBank mRNA AF109643.1 - 4 - 5 567 171
AF109644.1 - 4 - 5 567 171
BC088313.1 - 4 - 5 638 171

View Full Product Details