The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012231.1 | NP_001012231.1 | 1 - 1 | 395 | 124 |
XM_006251896.2 | XP_006251958.1 | 2 - 2 | 2533 | 124 |