Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NMS
Entrez Gene ID
497196
Gene Name
neuromedin S
Gene Aliases
-
Chromosome Location
Chr.9: 45605552 - 45616912 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012233.1 NP_001012233.1 4 - 5 212 108
GenBank mRNA AB164465.1 - 4 - 5 212 108

View Full Product Details