Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stfa2l1
Entrez Gene ID
408214
Gene Name
stefin A2-like 1
Gene Aliases
Stfa2
Chromosome Location
Chr.11: 67395889 - 67400201 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004129.1 NP_001004129.1 2 - 3 187 136

View Full Product Details