Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krt73
Entrez Gene ID
300248
Gene Name
keratin 73, type II
Gene Aliases
Kb36
Chromosome Location
Chr.7: 143399878 - 143408316 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008808.1 NP_001008808.1 6 - 7 1105 129
XM_008765703.1 XP_008763925.1 6 - 7 1145 129

View Full Product Details