The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022235.2 | NP_071571.1 | 2 - 2 | 957 | 174 |
GenBank mRNA | BC086406.1 | - | 2 - 2 | 957 | 174 |