Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rnase13
Entrez Gene ID
497194
Gene Name
ribonuclease, RNase A family, 13 (non-active)
Gene Aliases
-
Chromosome Location
Chr.15: 28322904 - 28325870 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012231.1 NP_001012231.1 1 - 1 311 71

View Full Product Details