Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
Raet1l
Entrez Gene ID
292461
Gene Name
retinoic acid early transcript 1L
Gene Aliases
-
Chromosome Location
Chr.7: 114233896 - 114234243 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013063.1 NP_001013081.1 - 949 79
GenBank mRNA BC079076.1 - - 949 79

View Full Product Details