Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pcdhb1
Entrez Gene ID
364841
Gene Name
protocadherin beta 1
Gene Aliases
-
Chromosome Location
Chr.18: 30282645 - 30298210 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001054930.4 XP_001054930.1 1 - 1 3361 83

View Full Product Details