Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pcdhb22
Entrez Gene ID
307486
Gene Name
protocadherin beta 22
Gene Aliases
pcdh-T5
Chromosome Location
Chr.18: 30592787 - 30595456 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001056235.5 XP_001056235.1 1 - 1 1122 155

View Full Product Details