The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024787.1 | NP_001019958.1 | 4 - 5 | 1088 | 112 |
XM_006244893.2 | XP_006244955.1 | 4 - 5 | 1137 | 112 | |
XM_006244894.2 | XP_006244956.1 | 4 - 5 | 1101 | 112 | |
GenBank mRNA | BC078974.1 | - | 4 - 5 | 1108 | 112 |
BC097950.1 | - | 4 - 5 | 1088 | 112 |