Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RT1-CE7
Entrez Gene ID
368153
Gene Name
RT1 class I, locus CE7
Gene Aliases
RT1-C/E7
Chromosome Location
Chr.20: 4695765 - 4698712 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008845.1 NP_001008845.1 6 - 7 1052 115

View Full Product Details