Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Epn2
Entrez Gene ID
60443
Gene Name
epsin 2
Gene Aliases
-
Chromosome Location
Chr.10: 47795496 - 47857338 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033914.1 NP_001029086.1 4 - 5 1138 71
NM_021852.2 NP_068624.2 3 - 4 967 71
XM_006246522.2 XP_006246584.1 5 - 6 1220 71
GenBank mRNA AF096269.1 - 2 - 3 708 71
BC094524.1 - 4 - 5 1117 71
FQ211574.1 - 5 - 6 1220 71
FQ211939.1 - 4 - 5 1162 71

View Full Product Details