Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lhb
Entrez Gene ID
25329
Gene Name
luteinizing hormone beta polypeptide
Gene Aliases
-
Chromosome Location
Chr.1: 101410021 - 101413725 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033975.1 NP_001029147.1 1 - 2 231 65
NM_012858.2 NP_036990.1 2 - 3 192 65
XM_006228975.2 XP_006229037.1 5 - 6 1997 65
GenBank mRNA CB566905.1 - - 195 65
CB812721.1 - 1 - 2 232 65
D00576.1 - 1 - 2 174 65
V01542.1 - 2 - 3 184 65

View Full Product Details