The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175604.2 | NP_783194.2 | 4 - 5 | 786 | 66 |
GenBank mRNA | BC088428.1 | - | 4 - 5 | 751 | 66 |
BC099797.1 | - | 4 - 5 | 786 | 66 |