The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034945.1 | NP_001030117.1 | 6 - 7 | 1065 | 83 |
XM_006249464.2 | XP_006249526.1 | 6 - 7 | 1082 | 83 | |
GenBank mRNA | BC103639.1 | - | 6 - 7 | 1065 | 83 |