Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spag11b
Entrez Gene ID
652998
Gene Name
sperm associated antigen 11b
Gene Aliases
Spag11c
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037852.1 NP_001032941.1 1 - 2 121 131
GenBank mRNA DQ012093.1 - 1 - 2 121 131

View Full Product Details