Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Defa24
Entrez Gene ID
290856
Gene Name
defensin alpha 24
Gene Aliases
Defcr24, Defcr4, Rd5, defensin 6, defensin 7, defensin-5
Chromosome Location
Chr.16: 75345086 - 75346122 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013053.1 NP_001013071.1 1 - 1 163 95

View Full Product Details