Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mfsd8
Entrez Gene ID
361939
Gene Name
major facilitator superfamily domain containing 8
Gene Aliases
Ab2-276, RGD1310132
Chromosome Location
Chr.2: 127708747 - 127784129 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047910.1 NP_001041375.1 6 - 7 791 120
GenBank mRNA AY325200.1 - 6 - 7 791 120

View Full Product Details