The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025624.2 | NP_001020795.1 | 5 - 6 | 393 | 103 |
GenBank mRNA | BC099178.1 | - | 5 - 6 | 359 | 103 |
BC099815.1 | - | 5 - 6 | 368 | 103 | |
CB614313.1 | - | 5 - 6 | 402 | 103 | |
FQ213300.1 | - | 5 - 6 | 361 | 103 | |
FQ213727.1 | - | 5 - 6 | 103 | ||
FQ214234.1 | - | 3 - 4 | 188 | 103 | |
FQ214394.1 | - | 5 - 6 | 352 | 103 | |
FQ214422.1 | - | 5 - 6 | 363 | 103 | |
FQ215102.1 | - | 5 - 6 | 369 | 103 | |
FQ216264.1 | - | 5 - 6 | 387 | 103 | |
FQ216489.1 | - | 5 - 6 | 324 | 103 | |
FQ217912.1 | - | 5 - 6 | 363 | 103 | |
FQ219745.1 | - | 5 - 6 | 428 | 103 | |
FQ219836.1 | - | 5 - 6 | 390 | 103 | |
FQ220601.1 | - | 5 - 6 | 404 | 103 | |
FQ221300.1 | - | 5 - 6 | 334 | 103 | |
FQ229179.1 | - | 5 - 6 | 352 | 103 | |
FQ229184.1 | - | 5 - 6 | 326 | 103 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_003751587.3 | XP_003751635.1 | 1 - 1 | 359 | 103 |