Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vom1r27
Entrez Gene ID
494297
Gene Name
vomeronasal 1 receptor 27
Gene Aliases
V1re19
Chromosome Location
Chr.1: 68528976 - 68529893 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008952.1 NP_001008952.1 1 - 1 733 105
GenBank mRNA AY510351.1 - 1 - 1 733 105

Gene Symbol
LOC100911644
Entrez Gene ID
100911644
Gene Name
vomeronasal type-1 receptor 4-like
Gene Aliases
-
Chromosome Location
Chr.1: 68649663 - 68650566 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008758880.1 XP_008757102.1 1 - 1 721 105

View Full Product Details