Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vom2r28
Entrez Gene ID
292547
Gene Name
vomeronasal 2 receptor, 28
Gene Aliases
RGD1565786
Chromosome Location
Chr.1: 69558008 - 69589096 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099472.1 NP_001092942.1 3 - 4 494 107

View Full Product Details