Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cttnbp2nl
Entrez Gene ID
310760
Gene Name
CTTNBP2 N-terminal like
Gene Aliases
RGD1310822
Chromosome Location
Chr.2: 207495056 - 207541642 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107712.2 NP_001101182.2 3 - 4 244 113
XM_006233087.1 XP_006233149.1 2 - 3 221 113
GenBank mRNA CB724536.1 - 3 - 4 262 113

View Full Product Details