The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009968.1 | NP_001009968.1 | 4 - 4 | 456 | 143 |
GenBank mRNA | AB180081.1 | - | 4 - 4 | 456 | 143 |