Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
Vom2r13
Entrez Gene ID
690593
Gene Name
vomeronasal 2 receptor, 13
Gene Aliases
-
Chromosome Location
Chr.1: 64928503 - 64975956 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099489.1 NP_001092959.1 - 1655 88

Gene Symbol
LOC100910184
Entrez Gene ID
100910184
Gene Name
vomeronasal type-2 receptor 26-like
Gene Aliases
-
Chromosome Location
Chr.11: 38674159 - 38717961 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008768625.1 XP_008766847.1 7 - 7 88

Gene Symbol
Vom2r12
Entrez Gene ID
690566
Gene Name
vomeronasal 2 receptor, 12
Gene Aliases
-
Chromosome Location
Chr.1: 64849657 - 64950897 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099488.1 NP_001092958.1 6 - 6 88

View Full Product Details