Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pck2
Entrez Gene ID
361042
Gene Name
phosphoenolpyruvate carboxykinase 2 (mitochondrial)
Gene Aliases
PEPCK-M
Chromosome Location
Chr.15: 34216735 - 34224357 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108377.2 NP_001101847.2 - 1114 71
GenBank mRNA BC166766.1 - - 1087 71

View Full Product Details