Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim43a
Entrez Gene ID
300884
Gene Name
tripartite motif-containing 43A
Gene Aliases
RGD1561074
Chromosome Location
Chr.8: 96389344 - 96401733 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142944.1 NP_001136416.1 1 - 2 411 118
XM_006243512.2 XP_006243574.1 2 - 3 651 118

View Full Product Details