The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135010.1 | NP_001128482.1 | 4 - 5 | 601 | 107 |
GenBank mRNA | FQ229939.1 | - | 4 - 5 | 663 | 107 |