Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ctdnep1
Entrez Gene ID
287447
Gene Name
CTD nuclear envelope phosphatase 1
Gene Aliases
Dullard
Chromosome Location
Chr.10: 56591261 - 56600235 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100494.1 NP_001093964.1 5 - 6 682 91
XM_006246643.2 XP_006246705.1 6 - 7 807 91
XM_006246644.2 XP_006246706.1 6 - 7 807 91
XM_008767804.1 XP_008766026.1 5 - 6 711 91
GenBank mRNA BC087638.1 - 3 - 4 316 91
BC107474.1 - 4 - 5 343 91

View Full Product Details