The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173118.2 | NP_775141.2 | 3 - 4 | 463 | 96 |
GenBank mRNA | AJ515237.1 | - | 3 - 4 | 398 | 96 |
BC058132.1 | - | 3 - 4 | 426 | 96 | |
FQ214040.1 | - | 3 - 4 | 384 | 96 | |
FQ216545.1 | - | 3 - 4 | 388 | 96 | |
FQ219301.1 | - | 3 - 4 | 405 | 96 | |
FQ219863.1 | - | 3 - 4 | 396 | 96 | |
FQ220660.1 | - | 3 - 4 | 384 | 96 | |
FQ220919.1 | - | 3 - 4 | 390 | 96 | |
FQ228456.1 | - | 3 - 4 | 388 | 96 | |
FQ229422.1 | - | 3 - 4 | 443 | 96 | |
FQ229782.1 | - | 3 - 4 | 384 | 96 | |
FQ229783.1 | - | 3 - 4 | 386 | 96 |