Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Rpl38
- Entrez Gene ID
-
689284
- Gene Name
- ribosomal protein L38
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 103198424 - 103202059 on Build Rnor_6.0
- Gene Symbol
- LOC685963
- Entrez Gene ID
-
685963
- Gene Name
- similar to 60S ribosomal protein L38
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 164002409 - 164002757 on Build Rnor_6.0
- Gene Symbol
- LOC690468
- Entrez Gene ID
-
690468
- Gene Name
- similar to 60S ribosomal protein L38
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 199478992 - 199479315 on Build Rnor_6.0
View Full Product Details