Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vom1r16
Entrez Gene ID
100312668
Gene Name
vomeronasal 1 receptor 16
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167549.1 NP_001161021.1 1 - 1 151 93

Gene Symbol
Vom1r111
Entrez Gene ID
494242
Gene Name
vomeronasal 1 receptor 111
Gene Aliases
V1rf4
Chromosome Location
Chr.1: 61219351 - 61220256 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009522.1 NP_001009522.1 1 - 1 151 93
GenBank mRNA AY510366.1 - 1 - 1 151 93

View Full Product Details