Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Mir18a
- Entrez Gene ID
-
100314227
- Gene Name
- microRNA 18a
- Gene Aliases
-
rno-mir-18a
- Chromosome Location
-
Chr.15: 100180015 - 100180110 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_031819.1 |
|
1 - 1 |
44 |
74 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 17 |
Mir17 |
Mir17-1, rno-mir-17-1 |
-53.0
|
Sense |
Chr. - 100179879 - 100179962 on Build Rnor_6.0 |
Downstream |
microRNA 19a |
Mir19a |
rno-mir-19a |
52.0
|
Sense |
Chr. - 100180162 - 100180243 on Build Rnor_6.0 |
View Full Product Details