Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Mir27a
- Entrez Gene ID
-
100314006
- Gene Name
- microRNA 27a
- Gene Aliases
-
rno-mir-27a
- Chromosome Location
-
Chr.19: 25318736 - 25318822 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_031833.1 |
|
1 - 1 |
42 |
62 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 23a |
Mir23a |
rno-mir-23a |
-80.0
|
Sense |
Chr. - 25318582 - 25318656 on Build Rnor_6.0 |
Downstream |
microRNA 24-2 |
Mir24-2 |
rno-mir-24-2 |
41.0
|
Sense |
Chr. - 25318863 - 25318970 on Build Rnor_6.0 |
View Full Product Details