Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnf24
Entrez Gene ID
362218
Gene Name
ring finger protein 24
Gene Aliases
-
Chromosome Location
Chr.3: 123848825 - 123849788 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108591.2 NP_001102061.1 1 - 2 41 62

View Full Product Details