Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dclk3
Entrez Gene ID
316023
Gene Name
doublecortin-like kinase 3
Gene Aliases
Dcamkl3, RGD1309232
Chromosome Location
Chr.8: 119691567 - 119744484 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191800.1 NP_001178729.1 3 - 4 2067 100

View Full Product Details